Skip to Content
Merck
All Photos(1)

Documents

75190

Sigma-Aldrich

Heptanoic acid

≥99.0% (GC)

Synonym(s):

Enanthic acid, Oenanthic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.5 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

expl. lim.

10.1 %

refractive index

n20/D 1.4221 (lit.)
n20/D 1.423

bp

223 °C (lit.)

mp

−10.5 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(O)=O

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heptanoic acid can be used:
  • For esterification with glycerol to synthesize a triacylglycerol called as triheptanoin or trienanthoin.
  • To synthesize 1-hexene via decarbonylation reaction by using platinum nanoparticles supported on carbon.
  • To synthesize 7-tridecanone by ketonization in the presence of MnO2/CeO2 or ZrO2 catalysts supported on the surface of alumina.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decarbonylation of heptanoic acid over carbon-supported platinum nanoparticles.
Lopez-Ruiz JA and Davis RJ
Green Chemistry, 16(2), 683-694 (2014)
Decarboxylative coupling of heptanoic acid. Manganese, cerium and zirconium oxides as catalysts.
Glinski M and Kijenski J
Applied Catalysis A: General, 190(1-2), 87-91 (2000)
Chemical synthesis of tricaproin, trienantin and tricaprylin.
da Rocha Ataide T, et al.
International Journal of Food Science and Technology, 42(12), 1504-1508 (2007)
Hafiz Umer Javed et al.
Food research international (Ottawa, Ont.), 107, 649-659 (2018-03-28)
The aroma profiles of volatile compounds (VOCs) were analyzed by GC/MS in pre-treated air-dried (PAD) and sun-dried (PSD) raisins during storage. Total 98, 94 and 81 VOCs were identified in Zixiang Seedless raisins (ZSRs), Centennial Seedless raisins (CSRs) and Thompson
Lara A Skelton et al.
Physiological reports, 3(3) (2015-03-18)
The renal proximal tubule (PT) plays a major role in whole-body pH homeostasis by secreting H(+) into the tubule lumen. Previous work demonstrated that PTs respond to basolateral changes in [CO2] and [HCO3-] by appropriately altering H(+) secretion-responses blocked by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service