Skip to Content
Merck
All Photos(1)

Documents

749737

Sigma-Aldrich

Indium tin oxide coated PET

surface resistivity 100 Ω/sq, L × W × thickness 1 ft × 1 ft × 7 mil, sheet

Synonym(s):

ITO-PET, Polyethylene terephthalate film, ITO coated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

sheet

surface resistivity

100 Ω/sq

L × W × thickness

1 ft × 1 ft × 7 mil

SMILES string

O=[In]O[In]=O.O=[Sn]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive. The ITO coating is protected by a thin, transparent film, that can be easily peeled off.

Application

ITO-PET may be used in fabricating flexible OLEDs. ITO/PET may be used as a reference anode in conventional flexible polymer based organic solar cells (OSCs).

Features and Benefits

The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.
These polyester film products comprise of sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of glass substrates.

Physical properties

Thickness of ITO coating is 1000 Å

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance Organic Light-Emitting Diodes Using ITO Anodes Grown on Plastic by Room-Temperature Ion-Assisted Deposition
Yang Y, et al.
Advanced Materials, 16(4), 321-324 (2004)
Modular mass spectrometric tool for analysis of composition and phosphorylation of protein complexes
Blethrow JD, et al.
PLoS ONE, 2(4), e358-e358 (2007)
Efficient and Flexible ITO-Free Organic Solar Cells Using Highly Conductive Polymer Anodes.
Na SI, et al.
Advanced Materials, 20(21), 4061-4067 (2008)

Articles

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service