745847
Vanadium(IV) carbide
powder, <2 μm, 99.9% trace metals basis
Synonym(s):
Vanadium monocarbide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
VC
CAS Number:
Molecular Weight:
62.95
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
powder
particle size
<2 μm
mp
2810 °C (lit.)
density
5.77 g/mL at 25 °C (lit.)
SMILES string
[C-]#[V+]
InChI
1S/C.V/q-1;+1
InChI key
ZLANVVMKMCTKMT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Vanadium Carbide has drawn research interest both as a potential component in anode materials for lithium ion batteries, and in supercapacitors , as well as serving as a coating material and an additive in ceramic composites. In addition, nitrogen doped vanadium carbide has been studied as a potential counter electrode catalyst in dye sensitized solar cells.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Li; M.Q.;
Electrochimica Acta, 54(19), 4506-4513 (2009)
Aghaie-Khafri; M.;
Journal of Physics and Chemistry of Solids, 69(10), 2465-2470 (2008)
Mingxing Wu et al.
Journal of the American Chemical Society, 134(7), 3419-3428 (2012-01-28)
Three classes (carbides, nitrides and oxides) of nanoscaled early-transition-metal catalysts have been proposed to replace the expensive Pt catalyst as counter electrodes (CEs) in dye-sensitized solar cells (DSCs). Of these catalysts, Cr(3)C(2), CrN, VC(N), VN, TiC, TiC(N), TiN, and V(2)O(3)
Thomberg; T.;
Journal of Electroanalytical Chemistry, 630(1-2), 55-62 (2009)
Yang; F. Z.;
Journal of Materials Processing Technology, 209(9), 4531-4536 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service