Skip to Content
Merck
All Photos(2)

Key Documents

740691

Sigma-Aldrich

3-(3-Hydroxypropyl)-1H-indole

97%

Synonym(s):

Homotryptophol, 1H-indole-3-proanol, 3-Indolepropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.605

density

1.147 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCCc1c[nH]c2ccccc12

InChI

1S/C11H13NO/c13-7-3-4-9-8-12-11-6-2-1-5-10(9)11/h1-2,5-6,8,12-13H,3-4,7H2

InChI key

LYPSVQXMCZIRGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikas Sadauskas et al.
Biomolecules, 10(4) (2020-04-30)
A plant auxin hormone indole-3-acetic acid (IAA) can be assimilated by bacteria as an energy and carbon source, although no degradation has been reported for indole-3-propionic acid and indole-3-butyric acid. While significant efforts have been made to decipher the Iac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service