Skip to Content
Merck
All Photos(2)

Key Documents

738581

Sigma-Aldrich

Platinum on graphitized carbon

greener alternative

extent of labeling: 10 wt. % loading

Synonym(s):

10% Pt on Vulcan XC72, 10% Pt/Vulcan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: platinum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

10 wt. % loading

particle size

<5 nm (Pt)

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on graphitized carbon (10% Pt/Vulcan XC72) is a high surface area material that is used as an electrocatalyst for the development of electrolytic fuel cells and direct methanol fuel cells. It can also be used in the formation of a non-enzymatic sensor for the oxidation of glucose.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of Cu2O/Carbon Vulcan XC-72 as non-enzymatic sensor for glucose determination
El Khatib KM and Hameed RMA
Biosensors And Bioelectronics, 26(8), 3542-3548 (2011)
Oxygen reduction on a high-surface area Pt/Vulcan carbon catalyst: a thin-film rotating ring-disk electrode study
Paulus UA, et al.
Journal of Electroanalytical Chemistry, 495(2), 134-145 (2001)
Load cycle durability of a graphitized carbon black-supported platinum catalyst in polymer electrolyte fuel cell cathodes
Takei C, et al.
Journal of Power Sources, 324, 729-737 (2016)
Platinum nanoparticles supported by a Vulcan XC-72 and PANI doped with trifluoromethane sulfonic acid substrate as a new electrocatalyst for direct methanol fuel cells
Gharibi H, et al.
The Journal of Physical Chemistry C, 114(11), 5233-5240 (2010)

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service