Skip to Content
Merck
All Photos(2)

Key Documents

738271

Sigma-Aldrich

3-Butynoic acid

95%

Synonym(s):

2-Ethynylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O2
CAS Number:
Molecular Weight:
84.07
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

Assay

95%

form

powder

mp

70-80 °C

SMILES string

OC(=O)CC#C

InChI

1S/C4H4O2/c1-2-3-4(5)6/h1H,3H2,(H,5,6)

InChI key

KKAHGSQLSTUDAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Some of the applications of 3-butynoic acid are:
  • Synthesis of functionalized γ-butyrolactones and tetrahydrofurans via Conia-ene cyclizations.
  • Synthesis of allenoates for [2+2] cycloadditions with alkenes.
  • Phosphine-catalyzed synthesis of highly functionalized coumarins.
  • Synthesis of various useful organotin reagents via radical hydrostannation of 3-butynoic acid.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic Diazo?OH Insertion/Conia?Ene Cascade Catalysis for the Stereoselective Synthesis of ??Butyrolactones and Tetrahydrofurans.
Hunter A C, et al.
Chemistry?A European Journal , 22(45), 16062-16065 (2016)
Catalytic Enantioselective Allenoate?Alkene [2+ 2] Cycloadditions.
Conner M L, et al.
Journal of the American Chemical Society, 137(10), 3482-3485 (2015)
Phosphine-catalyzed synthesis of highly functionalized coumarins.
Henry C E and Kwon O
Organic Letters, 9(16), 3069-3072 (2007)
Ionogels, new materials arising from the confinement of ionic liquids within silica-derived networks.
Neouze M A, et al.
Chemistry of Materials, 18(17), 3931-3936 (2006)
Zhipeng Li et al.
The Analyst, 145(12), 4239-4244 (2020-05-22)
The aim of this study was to overcome the reported shortcomings of the glutathione (GSH) detection of rhodamine-based fluorescent probes, such as poor selectivity to thiol groups and reversible unstable covalent binding with the thiol groups. Here, we have developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service