Skip to Content
Merck
All Photos(1)

Documents

737402

Sigma-Aldrich

Hexahydro-4-azepinone hydrochloride

96%

Synonym(s):

4-Azepanone hydrochloride, 4-Oxohomopiperidine hydrochloride, Hexahydro-4H-azepin-4-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO · HCl
CAS Number:
Molecular Weight:
149.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

176-181 °C

SMILES string

Cl.O=C1CCCNCC1

InChI

1S/C6H11NO.ClH/c8-6-2-1-4-7-5-3-6;/h7H,1-5H2;1H

InChI key

WFTRLIZPJMFJER-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service