Skip to Content
Merck
All Photos(1)

Key Documents

733679

Sigma-Aldrich

6-Amino-1-hexanethiol hydrochloride

Synonym(s):

6-Mercaptohexylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NS · HCl
CAS Number:
Molecular Weight:
169.72
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

125-130 °C

storage temp.

2-8°C

SMILES string

Cl.NCCCCCCS

InChI

1S/C6H15NS.ClH/c7-5-3-1-2-4-6-8;/h8H,1-7H2;1H

InChI key

BFCMUBZJTPADOD-UHFFFAOYSA-N

General description

6-Amino-1-hexanethiol hydrochloride(6-AHT) is an amine terminated alkanethiol which forms a self-assembled monolayer(SAM) on a variety of surfaces.

Application

6-AHT can be used in the surface modification of gold surface(Au) for the development of cell adhesive surfaces. It may also be used as a linker with quantum dots(QDs) which forms amine linkages with the carboxy groups of the fullerene heterodimers(FMH) which finds potential applications in molecular electronics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of surface roughness on self-assembled monolayer plasmonic ruler in nonlocal regime.
Hajisalem G, et al.
Optics Express, 22(8), 9604-9610 (2014)
Coulomb Blockade Plasmonic Switch.
Xiang D, et al.
Nano Letters, 17(4), 2584-2588 (2017)
Effect of surface potential on epithelial cell adhesion, proliferation and morphology.
Chang H, et al.
Colloids and Surfaces. B, Biointerfaces, 141(8), 179-186 (2016)
Kamila Malecka et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
The background: The monolayers self-assembled on the gold electrode incorporated transition metal complexes can act both as receptor ("host" molecules) immobilization sites, as well as transducer for interface recognitions of "guest" molecules present in the aqueous solutions. Their electrochemical parameters
Quantum dot-bridge-fullerene heterodimers with controlled photoinduced electron transfer.
Zhihua Xu et al.
Angewandte Chemie (International ed. in English), 50(27), 6079-6083 (2011-05-12)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service