Skip to Content
Merck
All Photos(1)

Documents

727954

Sigma-Aldrich

1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide

98%

Synonym(s):

HMIM BTI, HMIM TFS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19F6N3O4S2
CAS Number:
Molecular Weight:
447.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)
98%

form

liquid

SMILES string

CCCCCCn1cc[n+](C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C10H19N2.C2F6NO4S2/c1-3-4-5-6-7-12-9-8-11(2)10-12;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h8-10H,3-7H2,1-2H3;/q+1;-1

InChI key

RCNFOZUBFOFJKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide is a 1-alkyl-3-methylimidazolium based ionic liquid. The solubility of gases such as oxygen and methane in HMIM TFS can be improved in the presence of carbon dioxide.

Application

  • Electrochemical behavior and determination of rutin on modified carbon paste electrodes.: This research investigates the electrochemical properties of rutin using modified carbon paste electrodes incorporating 1-Hexyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, an ionic liquid. The study highlights the ionic liquid′s role in enhancing the electrode′s sensitivity and stability, making it a promising material for electrochemical sensing applications (Macikova et al., 2012).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluid-phase behavior of {1-hexyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide,[C6mim][NTf2],+ C2? C8 n-alcohol} mixtures: Liquid? liquid equilibrium and excess volumes
Lachwa, Joanna, et al.
Journal of Chemical and Engineering Data, 51.6, 2215-2221 (2006)
Excess properties of binary mixtures containing 1-hexyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide ionic liquid and polar organic compounds
Gonzalez, Emilio J., Angeles Dominguez, and Eugenia A. Macedo
The Journal of Chemical Thermodynamics, 47, 300-311 (2012)
Thermophysical properties of 1-hexyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide at high temperatures and pressures
Safarov, Javid, et al.
Journal of Molecular Liquids, 187, 137-156 (2013)
Surface tension measurements with validated accuracy for four 1-alkyl-3-methylimidazolium based ionic liquids.
Klomfar J, et al.
The Journal of Chemical Thermodynamics, 42(3), 323-329 (2010)
Low-temperature heat capacity of room-temperature ionic liquid, 1-hexyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide
Shimizu, Yoshitaka, et al.
The Journal of Physical Chemistry B, 110.28, 13970-13975 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service