Skip to Content
Merck
All Photos(5)

Key Documents

703478

Sigma-Aldrich

trans-2-Bromovinylboronic acid MIDA ester

technical grade

Synonym(s):

BB1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9BBrNO4
CAS Number:
Molecular Weight:
261.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

solid

functional group

bromo

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C\Br

InChI

1S/C7H9BBrNO4/c1-10-4-6(11)13-8(2-3-9)14-7(12)5-10/h2-3H,4-5H2,1H3/b3-2+

InChI key

ALXOFLXCBIRRRC-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Prielhofer et al.
BMC systems biology, 11(1), 123-123 (2017-12-10)
State-of-the-art strain engineering techniques for the host Pichia pastoris (syn. Komagataella spp.) include overexpression of homologous and heterologous genes, and deletion of host genes. For metabolic and cell engineering purposes the simultaneous overexpression of more than one gene would often
S Landmann et al.
The Journal of experimental medicine, 194(4), 379-391 (2001-08-22)
Cell surface expression of major histocompatibility complex class II (MHCII) molecules is increased during the maturation of dendritic cells (DCs). This enhances their ability to present antigen and activate naive CD4(+) T cells. In contrast to increased cell surface MHCII
Yuan-Lin Chou et al.
PLoS pathogens, 9(6), e1003405-e1003405 (2013-06-12)
The triple-gene-block protein 3 (TGBp3) of Bamboo mosaic virus (BaMV) is an integral endoplasmic reticulum (ER) membrane protein which is assumed to form a membrane complex to deliver the virus intracellularly. However, the virus entity that is delivered to plasmodesmata
Sarah Al-Maawi et al.
Acta biomaterialia, 121, 621-636 (2020-11-30)
Evaporation of phosphate species during thermal treatment (> 400 °C) of calcium phosphates leads to the formation of an alkaline layer on their surface. The aim of this study was to evaluate the hypothesis that the biological response of thermally treated
Bin-Zhi Qian et al.
The Journal of experimental medicine, 212(9), 1433-1448 (2015-08-12)
Although the link between inflammation and cancer initiation is well established, its role in metastatic diseases, the primary cause of cancer deaths, has been poorly explored. Our previous studies identified a population of metastasis-associated macrophages (MAMs) recruited to the lung

Articles

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service