Skip to Content
Merck
All Photos(1)

Key Documents

69868

Sigma-Aldrich

Magnesium monoperoxyphthalate hexahydrate

technical, ~80% (RT)

Synonym(s):

Magnesium bis(monoperoxyphthalate) hexahydrate, MMPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CC6H4CO3)2Mg·6H2O
Molecular Weight:
494.64
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

~80% (RT)

form

powder

reaction suitability

reagent type: oxidant

mp

93 °C (dec.) (lit.)

functional group

hydroperoxide

SMILES string

O.O.O.O.O.O.OOC(=O)c1ccccc1C(=O)O[Mg]OC(=O)c2ccccc2C(=O)OO

InChI

1S/2C8H6O5.Mg.6H2O/c2*9-7(10)5-3-1-2-4-6(5)8(11)13-12;;;;;;;/h2*1-4,12H,(H,9,10);;6*1H2/q;;+2;;;;;;/p-2

InChI key

WWOYCMCZTZTIGU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium monoperoxyphthalate hexahydrate is an organic peroxide used in many oxidation reactions. It exhibits non-deflagrating and non-shock-sensitive properties and also used as a surface disinfectant.

Application

Reagent for:
  • Trans-diaxial hydroxylation under bismuth(III) triflate catalyst
  • Oxidation rections under mild conditions
  • Epoxidation of unsaturated steroids
  • Synthesis of ring-B oxygenated steroids as anticancer agents

Other Notes

Versatile oxidant soluble in water and the lower alcohols.; Oxidations are carried out in homogenous solution (alcohol, aqueous alcohol) or in a two-phase system under PTC (chloroform-water)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Org. Perox. E - Skin Corr. 1C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel organic peroxygen reagents for use in organic synthesis
Organic peroxygen chemistry, 1-19 (1993)
D. Enders et al.
Synlett, 725-725 (1990)
F. Fringuelli et al.
The Journal of Organic Chemistry, 57, 1198-1198 (1992)
P. Brougham et al.
Synthesis, 1015-1015 (1987)
Efficacy of three surface disinfectants against spores of Clostridium difficile ribotype 027
Horejsh D and Kampf G
International Journal of Hygiene and Environmental Health, 214(2), 172-174 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service