Skip to Content
Merck
All Photos(1)

Key Documents

688886

Sigma-Aldrich

Methyl acetoacetate

Arxada quality, ≥99% (GC)

Synonym(s):

3-Oxobutanoic acid methyl ester, Acetoacetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH3
CAS Number:
Molecular Weight:
116.12
Beilstein:
506727
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

536 °F

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% acid (as acetic acid)
≤0.10% water

color

APHA: ≤20

refractive index

n20/D 1.419 (lit.)

bp

169-170 °C/70 mmHg (lit.)

mp

−80 °C (lit.)

density

1.076 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC(C)=O

InChI

1S/C5H8O3/c1-4(6)3-5(7)8-2/h3H2,1-2H3

InChI key

WRQNANDWMGAFTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(47), 16796-16801 (2014-11-05)
Much remains unknown regarding speciation. Host-pathogen interactions are a major driving force for diversification, but the genomic basis for speciation and host shifting remains unclear. The fungal genus Metarhizium contains species ranging from specialists with very narrow host ranges to
L Sweetman et al.
Biomedical mass spectrometry, 5(3), 198-207 (1978-03-01)
A number of previously unrecognized abnormal metabolites have been identified and quantitated in the urine of a patient with an inherited deficiency of propionyl-CoA carboxylase. These included the isoleucine metabolites 2-methyl-3-hydroxybutyric acid and 2-methylacetoacetic acid. These isomers 3-hydroxyvaleric acid and
Karl Sörelius et al.
Circulation, 130(24), 2136-2142 (2014-11-08)
Mycotic aortic aneurysm (MAA) is a rare and life-threatening disease. The aim of this European multicenter collaboration was to study the durability of endovascular aortic repair (EVAR) of MAA, by assessing late infection-related complications and long-term survival. All EVAR treated
S Aramaki et al.
Journal of inherited metabolic disease, 14(1), 63-74 (1991-01-01)
The concentrations of 2-methylacetoacetate, 2-methyl-3-hydroxybutyrate and tiglylglycine were determined by gas chromatography-mass spectrometry in urine collected before and for 8 h after loading with 100 mg of isoleucine per kg of body weight. The sum of 2-methylacetoacetate and 2-butanone, a
B H Robinson et al.
The Journal of pediatrics, 95(2), 228-233 (1979-08-01)
A female child presented at one year of age with a febrile illness and loose stools, then developed severe ketoacidosis with vomiting; an apparent salicylate level of 11 mg/dl was measured. A sibling had died in similar circumstances nine years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service