Skip to Content
Merck
All Photos(1)

Documents

687553

Sigma-Aldrich

Hydroxymethyl EDOT

95%

Synonym(s):

(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol, EDT-methanol, Thieno[3,4-b]-1,4-dioxin-2-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3S
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

42-46 °C

storage temp.

2-8°C

SMILES string

OCC1COc2cscc2O1

InChI

1S/C7H8O3S/c8-1-5-2-9-6-3-11-4-7(6)10-5/h3-5,8H,1-2H2

InChI key

YFCHAINVYLQVBG-UHFFFAOYSA-N

General description

Hydroxymethyl EDOT (EDT-methanol) is a conjugated polymer that is used as a precursor of ethylenedioxythiophene (EDOT). The hydroxymethyl groups in the EDOT monomers enhance the electro-polymerization in an aqueous solution to form an electro-active hydrophilic polymer.

Application

EDOT derivative useful in preparation of functional electroactive polymers
EDT-methanol can functionalize poly(L-lactic acid) by using organometallic polymerization, which can be used to form biodegradable and conducting macromonomers for biomedical applications. It can be polymerized to form poly(hydroxymethyl EDOT) based films, which may be incorporated with silver nanoparticles to form nanocomposites on polyethylene terephthalate (PET) for flexible plastic devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akoudad, S.; Roncali, J.
Electrochemical Communications, 2, 72-72 (2000)
Electrogenerated conjugated polymers incorporating a ferrocene-derivatized-(3, 4-ethylenedioxythiophene).
Brisset H, et al.
Electrochemical Communications, 6(3), 249-253 (2004)
Preparation of poly (hydroxymethyl EDOT)/nano-silver composite films by oxidative polymerization with low-baking temperature, low resistance and good adhesion on PET substrate.
Hsiao A, et al
Synthetic Metals, 160(21-22), 2319-2322 (2010)
Modification of the electrochemical and electronic properties of electrogenerated poly (3, 4-ethylenedioxythiophene) by hydroxymethyl and oligo (oxyethylene) substituents
Akoudad S and Roncali J
Electrochemical Communications, 2(1), 72-76 (2000)
Antonio Dominguez-Alfaro et al.
Polymers, 13(3) (2021-02-13)
Carbon nanomaterials (CNMs) and conjugated polymers (CPs) are actively investigated in applications such as optics, catalysis, solar cells, and tissue engineering. Generally, CNMs are implemented in devices where the relationship between the active elements and the micro and nanostructure has

Articles

Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service