All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H15NSi
CAS Number:
Molecular Weight:
189.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
form
solid
mp
94-98 °C
storage temp.
2-8°C
SMILES string
C[Si](C)(C)C#Cc1ccc(N)cc1
InChI
1S/C11H15NSi/c1-13(2,3)9-8-10-4-6-11(12)7-5-10/h4-7H,12H2,1-3H3
InChI key
CHTZIDLXGXGNMA-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Baybars Köksoy et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 147, 111886-111886 (2020-11-29)
The present work describes the first synthesis of novel asymmetric zinc (II) phthalocyanine (ZnPc) including three boron dipyrromethene (BODIPY) and one ethyloxy azido moieties. Moreover, single walled carbon nanotube (SWCNT) surface was functionalized by this ZnPc containing BODIPY; using the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service