Skip to Content
Merck
All Photos(1)

Key Documents

681539

Sigma-Aldrich

(S)-(–)-3-3′-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1′-bi-2-naphthol

95%

Synonym(s):

(1S)-3,3′-Bis[3,5-bis(trifluoromethyl)phenyl]1,1′-binaphthalene-2,2′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H18F12O2
CAS Number:
Molecular Weight:
710.51
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

(Powder or Crystals or Solid or Chunks)

optical activity

[α]22/D -56°, c = 1 in chloroform

mp

215-219 °C

functional group

fluoro

SMILES string

Oc1c(cc2ccccc2c1-c3c(O)c(cc4ccccc34)-c5cc(cc(c5)C(F)(F)F)C(F)(F)F)-c6cc(cc(c6)C(F)(F)F)C(F)(F)F

InChI

1S/C36H18F12O2/c37-33(38,39)21-9-19(10-22(15-21)34(40,41)42)27-13-17-5-1-3-7-25(17)29(31(27)49)30-26-8-4-2-6-18(26)14-28(32(30)50)20-11-23(35(43,44)45)16-24(12-20)36(46,47)48/h1-16,49-50H

InChI key

PGXMYJSTCAQJBY-UHFFFAOYSA-N

Application

(S)-(-)-3-3′-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1′-bi-2-naphthol reacts with tin(IV) chloride to form a Sn(IV) aryloxide Lewis acid, which can catalyze asymmetric Diels-Alder reactions.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of chiral tin (IV) aryloxide as a mild Lewis acid catalyst for enantioselective Diels-Alder reaction.
Kano T, et al.
Tetrahedron Letters, 47(6), 873-875 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service