Skip to Content
Merck
All Photos(4)

Key Documents

676578

Sigma-Aldrich

Bis(tri-tert-butylphosphine)palladium(0)

greener alternative

Synonym(s):

Pd(t-Bu3P)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H54P2Pd
CAS Number:
Molecular Weight:
511.05
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

258-272 °C

greener alternative category

storage temp.

−20°C

SMILES string

[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/2C12H27P.Pd/c2*1-10(2,3)13(11(4,5)6)12(7,8)9;/h2*1-9H3;

InChI key

MXQOYLRVSVOCQT-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

  • Catalyst for Suzuki coupling on a multisubstituted sp3-carbon (eq. 1)
  • Catalyst for Stille coupling reaction of aryl chlorides (eq. 2)
  • Catalyst for Negishi coupling reaction (eq. 3)
  • Catalyst for Heck coupling to form tetrasubstituted olefins (eq. 4)
  • Catalyst for Buchwald-Hartwig amination of aryl halide (eq. 5)
  • Catalyst for carbonylation of aryl halides with carbamoylsilanes (eq. 6)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichiro Itami et al.
Journal of the American Chemical Society, 126(38), 11778-11779 (2004-09-24)
We have developed a programmable and diversity-oriented synthetic scheme for tetrasubstituted olefins through a site-selective and sequential assembly of pi-components onto a C=C core of vinyl 2-pyrimidyl sulfide. Noteworthy features are that (i) all components assembled stem from readily available
Ryoichi Kuwano et al.
The Journal of organic chemistry, 67(18), 6479-6486 (2002-08-31)
The amination of aryl halides in the presence of inexpensive and air-stable alkali metal hydroxide bases and Pd[P(t-Bu)3]2 as catalyst gave arylamines in high yields. The reactions were conducted with a catalytic amount of cetyltrimethylammonium bromide as phase-transfer agent and
Robert F Cunico et al.
Organic letters, 5(26), 4947-4949 (2003-12-20)
Alkenyl chlorides and bromides are converted into tertiary enamides by treatment with a carbamoylsilane in toluene at 110 degrees C in the presence of phosphine-palladium(0) catalysts. [reaction: see text]
Kohei Endo et al.
Journal of the American Chemical Society, 132(32), 11033-11035 (2010-08-12)
The palladium-catalyzed Suzuki-Miyaura cross-coupling on a multisubstituted sp(3)-carbon in 1,1-diborylalkanes was achieved at room temperature. The generation of a monoborate intermediate by virtue of the adjacent B atom could result in the chemoselective coupling reaction under ambient conditions.
Negishi, E.-I.; Shi, J.-C.; Zeng, X.
Tetrahedron, 61, 9886-9886 (2005)

Articles

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service