Skip to Content
Merck
All Photos(1)

Key Documents

674389

Sigma-Aldrich

Phenylphosphine

ca. 10% weight in hexanes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5PH2
CAS Number:
Molecular Weight:
110.09
Beilstein:
741946
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.79 (vs air)

vapor pressure

760 mmHg ( 160 °C)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reagent type: ligand

refractive index

n20/D 1.578 (lit.)

bp

160 °C (lit.)

density

0.6912 g/mL at 25 °C
1.001 g/mL at 25 °C (lit.)

functional group

phosphine

storage temp.

−20°C

SMILES string

Pc1ccccc1

InChI

1S/C6H7P/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

RPGWZZNNEUHDAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongzhou Gu et al.
Nature, 465(7295), 202-205 (2010-05-14)
Our ability to synthesize nanometre-scale chemical species, such as nanoparticles with desired shapes and compositions, offers the exciting prospect of generating new functional materials and devices by combining them in a controlled fashion into larger structures. Self-assembly can achieve this
Nuli Xie et al.
ACS nano, 13(4), 4174-4182 (2019-04-05)
It is of great interest to construct DNA-functionalized gold nanoparticles (DNA-AuNPs) with a controllable number of DNA strands and relative orientations. Herein, we describe a three-dimensional (3D) molecular transfer strategy, in which a pattern of DNA strands can be transferred
Battulga Munkhbat et al.
ACS nano, 14(1), 1196-1206 (2020-01-07)
Hybrid light-matter states-polaritons-have attracted considerable scientific interest recently, motivated by their potential for development of nonlinear and quantum optical schemes. To realize such states, monolayer transition metal dichalcogenides (TMDCs) have been widely employed as excitonic materials. In addition to neutral
Ahibur Rahaman et al.
Dalton transactions (Cambridge, England : 2003), 46(10), 3207-3222 (2017-02-22)
The influence of the substitution, orientation and structure of the phosphido bridges in [Fe
Jiefang Sun et al.
Analytical chemistry, 91(10), 6454-6461 (2019-04-18)
Ricin is a highly toxic protein largely existing in castor beans, which could be used as a warfare agent due to its unique properties. As a deadenylase, inactivation of ricin means a loss of its toxic threat. Therefore, developing simple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service