All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H8F3N
CAS Number:
Molecular Weight:
139.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.379
bp
104-106 °C
density
1.210 g/mL at 25 °C
functional group
fluoro
storage temp.
2-8°C
SMILES string
FC(F)(F)C1CCCN1
InChI
1S/C5H8F3N/c6-5(7,8)4-2-1-3-9-4/h4,9H,1-3H2
InChI key
VINAMCOZNJHNIH-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
78.8 °F
Flash Point(C)
26 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service