Skip to Content
Merck
All Photos(1)

Key Documents

658057

Sigma-Aldrich

L-Cysteine-13C3,15N

≥99 atom %, ≥98% (CP)

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid-13C3,15N, 13C and 15N Labeled L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS13CH213CH(15NH2)13COOH
CAS Number:
Molecular Weight:
125.13
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥99 atom %

Assay

≥98% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

220 °C (dec.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

[H][13C@]([15NH2])([13CH2]S)[13C](O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1/i1+1,2+1,3+1,4+1

InChI key

XUJNEKJLAYXESH-UVYXLFMMSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan-Wilm Lackmann et al.
PloS one, 14(5), e0216606-e0216606 (2019-05-09)
Thiol moieties are major targets for cold plasma-derived nitrogen and oxygen species, making CAPs convenient tools to modulate redox-signaling pathways in cells and tissues. The underlying biochemical pathways are currently under investigation but especially the role of CAP derived RNS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service