Skip to Content
Merck
All Photos(1)

Key Documents

648701

Sigma-Aldrich

4-(2-Aminoethyl)pyridine

96%

Synonym(s):

2-(γ-Pyridyl)ethylamine, 2-(4-Pyridinyl)ethylamine, 2-(4-Pyridyl)ethanamine, 2-(4-Pyridyl)ethylamine, 2-(Pyridin-4-yl)ethan-1-amine, 2-(Pyridin-4-yl)ethanamine, 2-(Pyridine-4-yl)ethanamine, 4-(β-Aminoethyl)pyridine, 4-Pyridylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.5380 (lit.)

bp

104 °C/9 mmHg (lit.)

density

1.0238 g/mL at 25 °C (lit.)

SMILES string

NCCc1ccncc1

InChI

1S/C7H10N2/c8-4-1-7-2-5-9-6-3-7/h2-3,5-6H,1,4,8H2

InChI key

IDLHTECVNDEOIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willem M Albers et al.
International journal of molecular sciences, 14(2), 3500-3513 (2013-02-08)
Pyridyl derivatives of lipoic acid were prepared as ligands for the study of the interaction with thyroxine (T4). Thin self-assembled films of the ligands were prepared in 70% ethanol on gold and their interaction with T4 was studied by titration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service