Skip to Content
Merck

640468

4′-(4-Bromophenyl)acetophenone

97%

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C14H11BrO
CAS Number:
Molecular Weight:
275.14
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
227-236-9
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

UUVKNCRMWPNBNM-UHFFFAOYSA-N

InChI

1S/C14H11BrO/c1-10(16)11-2-4-12(5-3-11)13-6-8-14(15)9-7-13/h2-9H,1H3

SMILES string

CC(=O)c1ccc(cc1)-c2ccc(Br)cc2

assay

97%

form

solid

mp

129-133 °C (lit.)

functional group

bromo, ketone

Quality Level

Related Categories

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service