Skip to Content
Merck
All Photos(3)

Key Documents

637351

Sigma-Aldrich

Carbon nanotube, double-walled

50-80% carbon basis, O.D. × I.D. × L 5 nm × 1.3-2.0 nm × 50 μm

Synonym(s):

DWNT, Double wall carbon nanotubes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

50-80% carbon basis

form

powder

composition

carbon, >90% (trace metal basis)

O.D. × I.D. × L

5 nm × 1.3-2.0 nm × 50 μm

surface area

>600 m2/g

bulk density

0.12‑0.14 g/mL

SMILES string

[C]

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Structure of double walled carbon nanotubes (DWNTs) can be imagined as a two coaxial concentric single walled carbon nanotubes (SWNTs). It has been prepared by catalytic chemical vapor deposition. DWNTs exhibit excellent field emission properties and may be potentially useful as field emission displays (FEDs) and field emission transistors (FETs).[1][2]

Application

Carbon nanotube, double walled can be used for a variety of applications such as solar cells,[3] nanomechanical resonators,[4] film heaters,[5] biosensors,[6] and field emission displays.[7]

Packaging

Packaged in glass bottles

Preparation Note

Chemical Vapor Deposition (CVD) Method

Analysis Note

50-80% DWNT content, 10-40% other CNTs, remainder being amorphous carbon and residual metal catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational behaviors of multiwalled-carbon-nanotube-based nanomechanical resonators
Li C and Chou T
Applied Physics Letters, 84(1), 121-123 (2004)
Synthesis of single-and double-walled carbon nanotube forests on conducting metal foils
Hiraoka T, et al.
Journal of the American Chemical Society, 128(41), 13338-13339 (2006)
Double-walled carbon nanotube solar cells
Wei J, et al.
Nano Letters, 7(8), 2317-2321 (2007)
Preparation of large-area double-walled carbon nanotube films and application as film heater
Wu ZP and Wang JN
Physica E: Low-Dimensional Systems and Nanostructures, 42(1), 77-81 (2009)
Double-walled carbon nanotube based carbon paste electrode as xanthine biosensor
Anik U and Serdar C
Microchimica Acta, 166(3-4), 209-213 (2009)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service