Skip to Content
Merck
All Photos(1)

Documents

635057

Sigma-Aldrich

Titanium(IV) oxide, rutile

<001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm

Synonym(s):

Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

Quality Level

description

single side polished

Assay

≥99.9% trace metals basis

form

(single crystal substrate)
(single side polished)

hardness

7 mohs

L × W × thickness

10 mm × 10 mm × 0.5 mm

transmittance

0.5-4.5 μm

mp

1840 °C

density

4.17 g/mL at 25 °C (lit.)
4.26 g/mL at 25 °C

semiconductor properties

<001>

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Possesses improved photocatalytic activity.

Physical properties

Specific heat capacity: 0.17 (25°C) Cal. / (g. Deg); Linear expansion coefficient: a: 7.14 x 10-6, c: 9.19 x 10-6; Thermal optical coefficient: dη/dT: a: -0.72 x 10-6/K, c: -0.42 x 10-6/K

Physical form

tetragonal (a = 4.5936 Å, c = 2.9582 Å)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciano Gama Braga et al.
Cell reports, 33(7), 108397-108397 (2020-11-19)
The balance of phospho-signaling at the outer kinetochore is critical for forming accurate attachments between kinetochores and the mitotic spindle and timely exit from mitosis. A major player in determining this balance is the PP2A-B56 phosphatase, which is recruited to
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:
D Minetto et al.
Environment international, 66, 18-27 (2014-02-11)
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great
Roberta Tassinari et al.
Nanotoxicology, 8(6), 654-662 (2013-07-10)
The study explored possible reproductive and endocrine effects of short-term (5 days) oral exposure to anatase TiO2 nanoparticles (0, 1, 2 mg/kg body weight per day) in rat. Nanoparticles were characterised by scanning electron microscopy (SEM) and transmission electron microscopy
Elisa Moschini et al.
Toxicology letters, 222(2), 102-116 (2013-08-03)
Metal oxide NPs are abundantly produced in nanotech industries and are emitted in several combustion processes, suggesting the need to characterize their toxic impact on the human respiratory system. The acute toxicity and the morphological changes induced by copper oxide

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service