Skip to Content
Merck
All Photos(1)

Key Documents

608777

Sigma-Aldrich

Ammonia-14N

99.99 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
14NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

15N depleted

isotopic purity

99.99 atom % 14N

form

gas

bp

-33 °C (lit.)

mp

-78 °C (lit.)

mass shift

depleted

SMILES string

[14NH3]

InChI

1S/H3N/h1H3/i1+0

InChI key

QGZKDVFQNNGYKY-IGMARMGPSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Gas 2 - Press. Gas Liquefied gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Vallet et al.
Kidney international, 88(1), 137-145 (2015-03-12)
Recent studies suggest that alkalinizing treatments improve the course of chronic kidney disease (CKD), even in patients without overt metabolic acidosis. Here, we tested whether a decreased ability in excreting urinary acid rather than overt metabolic acidosis may be deleterious
Yvonne E Thomassen et al.
Vaccine, 32(24), 2782-2788 (2014-03-04)
Vero cells were grown adherent to microcarriers (Cytodex 1; 3 g L(-1)) using animal component free media in stirred-tank type bioreactors. Different strategies for media refreshment, daily media replacement (semi-batch), continuous media replacement (perfusion) and recirculation of media, were compared
Marilyn Huckans et al.
Brain and behavior, 4(2), 123-142 (2014-04-01)
BackgroundThe purpose of this study was to characterize hepatitis C virus (HCV)-associated differences in the expression of 47 inflammatory factors and to evaluate the potential role of peripheral immune activation in HCV-associated neuropsychiatric symptoms-depression, anxiety, fatigue, and pain. An additional
Yan Chen et al.
Biomaterials, 60, 111-120 (2015-05-20)
A novel multifunctional envelope-type mesoporous silica nanoparticle (MEMSN) system combining the merits of pH-responsiveness, non-toxicity and biological specificity, is demonstrated for drug delivery and magnetic resonance imaging (MRI). This system is constructed by immobilizing acetals on the surface of mesoporous
Emma Petiot et al.
Vaccine, 33(44), 5974-5981 (2015-06-16)
Mammalian cell cultures are increasingly used for the production of complex biopharmaceuticals including viral vectors and vaccines. HEK293 is the predominant cell line used for the transient expression of recombinant proteins and a well-established system for the production of viral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service