Skip to Content
Merck
All Photos(1)

Key Documents

606758

Sigma-Aldrich

Formaldehyde-12C solution

20% in H2O, 99.9 atom % 12C

Synonym(s):

Formalin-12C solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H12CHO
Molecular Weight:
30.02
UNSPSC Code:
12142200
PubChem Substance ID:

description

99.9 atom % 12C

isotopic purity

99.9 atom % 12C

concentration

20% in H2O
20% in H2O

mass shift

depleted

SMILES string

[12CH2]=O

InChI

1S/CH2O/c1-2/h1H2/i1+0

InChI key

WSFSSNUMVMOOMR-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility
Silvia Surinova et al.
EMBO molecular medicine, 7(9), 1166-1178 (2015-08-09)
Non-invasive detection of colorectal cancer with blood-based markers is a critical clinical need. Here we describe a phased mass spectrometry-based approach for the discovery, screening, and validation of circulating protein biomarkers with diagnostic value. Initially, we profiled human primary tumor
James Larkin et al.
The New England journal of medicine, 373(1), 23-34 (2015-06-02)
Nivolumab (a programmed death 1 [PD-1] checkpoint inhibitor) and ipilimumab (a cytotoxic T-lymphocyte-associated antigen 4 [CTLA-4] checkpoint inhibitor) have been shown to have complementary activity in metastatic melanoma. In this randomized, double-blind, phase 3 study, nivolumab alone or nivolumab plus
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
Richard Brad Jones et al.
PLoS pathogens, 10(8), e1004287-e1004287 (2014-08-15)
Resting memory CD4+ T-cells harboring latent HIV proviruses represent a critical barrier to viral eradication. Histone deacetylase inhibitors (HDACis), such as suberanilohydroxamic acid (SAHA), romidepsin, and panobinostat have been shown to induce HIV expression in these resting cells. Recently, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service