Skip to Content
Merck
All Photos(3)

Key Documents

594539

Sigma-Aldrich

4-Methoxycarbonylphenylboronic acid

≥95%

Synonym(s):

(4-Carbomethoxyphenyl)boronic acid, 4-Carbomethoxybenzeneboronic acid, 4-Methoxycarbonylbenzeneboronic acid, 4-borono-benzoic acid 1-methyl ester, p-(Methoxycarbonyl)boronic acid, p-(Methoxycarbonyl)phenylboronic acid, p-borono-benzoic acid methyl ester, Methyl 4-boronobenzoate, Methyl p-boronobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BO4
CAS Number:
Molecular Weight:
179.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

197-200 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(cc1)B(O)O

InChI

1S/C8H9BO4/c1-13-8(10)6-2-4-7(5-3-6)9(11)12/h2-5,11-12H,1H3

InChI key

PQCXFUXRTRESBD-UHFFFAOYSA-N

Application

Reagent used for
  • Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence
  • Copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides
  • One-pot ipso-nitration of arylboronic acids
  • Copper-catalyzed nitration
  • Cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling


  • Reagent used in Preparation of
  • Biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid
  • Chromenones and their bradykinin B1 antagonistic activit
  • Pt nanoparticles@Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injectio
  • Salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor

Other Notes

Contains varying amount of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of salicylate-based compounds as a novel class of methionine aminopeptidase inhibitors
Wang, W-L.; Chai, S. C.; Ye, Q-Z.
Bioorganic & Medicinal Chemistry, 21, 7151-7154 (2011)
Cheng Wang et al.
Journal of the American Chemical Society, 134(17), 7211-7214 (2012-04-11)
Pt nanoparticles of 2-3 nm and 5-6 nm in diameter were loaded into stable, porous, and phosphorescent metal-organic frameworks (MOFs 1 and 2) built from [Ir(ppy)(2)(bpy)](+)-derived dicarboxylate ligands (L(1) and L(2)) and Zr(6)(μ(3)-O)(4)(μ(3)-OH)(4)(carboxylate)(12) secondary building units, via MOF-mediated photoreduction of
Chromenones as potent bradykinin B1 antagonists
Bryan, M. C.; et al.
Bioorganic & Medicinal Chemistry, 22, 619-622 (2012)
A Highly Practical and Reliable Nickel Catalyst for Suzuki-Miyaura Coupling of Aryl Halides
Zhao, Y-L.; et al.
Advanced Synthesis & Catalysis, 353, 2881-2881 (2011)
Hao Yang et al.
Journal of the American Chemical Society, 129(5), 1132-1140 (2007-02-01)
alpha-Amino acid thiol esters derived from N-protected mono-, di-, and tripeptides couple with aryl, pi-electron-rich heteroaryl, or alkenyl boronic acids in the presence of stoichiometric Cu(I) thiophene-2-carboxylate and catalytic Pd(2)(dba)(3)/triethylphosphite to generate the corresponding N-protected peptidyl ketones in good-to-excellent yields

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service