Skip to Content
Merck
All Photos(1)

Documents

588695

Sigma-Aldrich

L-Asparagine-13C4 monohydrate

98 atom % 13C, 95% (CP)

Synonym(s):

(S)-2-Aminosuccinic acid 4-amide-13C4 monohydrate, (S)·-·(+)·-·2-·Aminosuccinamic acid-13C4, (S)-2-Aminosuccinic acid 4-amide-13C4 monohydrate, L-Aspartic acid 4-amide-13C4 monohydrate, 13C Labeled asparagine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N13CO13CH213CH(NH2)13CO2H · H2O
CAS Number:
Molecular Weight:
154.10
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 13C

Quality Level

Assay

95% (CP)

form

solid

optical activity

[α]25/D +31.5°, c = 1 in 1 M HCl

mp

233-235 °C (lit.)

mass shift

M+4

SMILES string

[H]O[H].N[13C@@H]([13CH2][13C](N)=O)[13C](O)=O

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1/i1+1,2+1,3+1,4+1;

InChI key

RBMGJIZCEWRQES-KFYBRNFPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service