Skip to Content
Merck
All Photos(1)

Key Documents

58790

Sigma-Aldrich

(+)-Potassium Ds-threo-isocitrate monobasic

≥98.0% (NT)

Synonym(s):

(1R,2S)-1-Hydroxy-1,2,3-propanetricarboxylic acid monopotassium salt, Ds-(+)-threo-Isocitric acid monopotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7KO7
CAS Number:
Molecular Weight:
230.21
Beilstein:
3767339
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (NT)

optical activity

[α]20/D +20.5±1°, c = 2% in H2O

mp

~185 °C (dec.)

functional group

carboxylic acid
hydroxyl

SMILES string

[K+].O[C@H]([C@H](CC(O)=O)C([O-])=O)C(O)=O

InChI

1S/C6H8O7.K/c7-3(8)1-2(5(10)11)4(9)6(12)13;/h2,4,9H,1H2,(H,7,8)(H,10,11)(H,12,13);/q;+1/p-1/t2-,4+;/m0./s1

InChI key

IVLPTBJBFVJENN-LEJBHHMKSA-M

Application

(+)-Potassium Ds-threo-isocitrate monobasic may be used in the assessment of isocitrate dehydrogenase 2 (Idh2) activity by Kornberg method.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Substrate for the characterization of isocitrate dehydrogenase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Eguchi et al.
Biochimica et biophysica acta, 990(2), 133-137 (1989-02-24)
NADP-dependent isocitrate dehydrogenase (EC 1.1.1.42) was purified to electrophoretic homogeneity from an extremely thermophilic bacterium, Thermus thermophilus HB8, and shown to be a dimeric protein of molecular weight 115,000, with a pI of 5.5. The amino acid composition of the
Sarah E Anderson et al.
The Analyst, 145(11), 3899-3908 (2020-04-17)
The enzyme isocitrate dehydrogenase 1 (IDH1) catalyzes the conversion of isocitrate to alpha-ketoglutarate (αKG) and has emerged as an important therapeutic target for glioblastoma multiforme (GBM). Current methods for assaying IDH1 remain poorly suited for high-throughput screening of IDH1 antagonists.
Sequential actions of SIRT1-RELB-SIRT3 coordinate nuclear-mitochondrial communication during immunometabolic adaptation to acute inflammation and sepsis.
Liu TF, et al.
The Journal of Biological Chemistry, 290(1), 396-408 (2015)
null
Ashley S Williams et al.
Cell metabolism, 31(1), 131-147 (2019-12-10)
This study sought to examine the functional significance of mitochondrial protein acetylation using a double knockout (DKO) mouse model harboring muscle-specific deficits in acetyl-CoA buffering and lysine deacetylation, due to genetic ablation of carnitine acetyltransferase and Sirtuin 3, respectively. DKO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service