Skip to Content
Merck
All Photos(3)

Key Documents

586617

Sigma-Aldrich

Acrylamide-13C3

99 atom % 13C, 98% (CP)

Synonym(s):

2-Propenamide-1,2,3-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H213C=13CH13CONH2
CAS Number:
Molecular Weight:
74.06
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

98% (CP)

technique(s)

protein expression: suitable

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

mass shift

M+3

SMILES string

N[13C](=O)[13CH]=[13CH2]

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1+1,2+1,3+1

InChI key

HRPVXLWXLXDGHG-VMIGTVKRSA-N

General description

Acrylamide-13C3 is an isotopic analogue acrylamide.

Application

Acrylamide-13C3 is used
to identify the formation of acrylamide in various food stuffs.
as an internal standard for the quantitative analysis of acrylamide in food samples using single stage LC/MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service