Skip to Content
Merck
All Photos(3)

Documents

578797

Sigma-Aldrich

3-Ethynylthiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4S
CAS Number:
Molecular Weight:
108.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5800 (lit.)

bp

152-153 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

C#Cc1ccsc1

InChI

1S/C6H4S/c1-2-6-3-4-7-5-6/h1,3-5H

InChI key

MJHLPKWONJUCFK-UHFFFAOYSA-N

Application

3-Ethynylthiophene may be used in the synthesis of the following:
  • 1-(2-bromobenzyl)-4-(thiophen-3-yl)-1H-1,2,3-triazole which is obtained by heating with 2-iodophenylethylazide in the presence of copper acetate monohydrate catalyst in N-methyl-2-pyrrolidone
  • N-benzyl-1-phenyl-5-(thiophen-3-yl)-4-pentyn-2-amine via a multi-step reaction process
  • [(C4H3S-3)C≡CAg]n, a polymeric compound obtained via reaction with silver nitrate in the presence of triethylamine in acetonitrile
  • 4,5-bis(thiophen-3-ylethynyl)phthalonitrile via Sonogashira cross-coupling reaction with 4,5-dichlorophthalonitrile

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Silver (I)- Thiophene p Interaction in the Assembly of Coordination Networks with the Supramolecular Synthons R- C? C? Ag n (R= 2-or 3-thienyl; n= 4)"
Zhao L, et al.
Organometallics, 26(18), 4439- 4448 (2007)
"Synthesis and photophysical properties of novel unsymmetrical metal-free and metallophthalocyanines"
Ozcesmeci I, et al.
Journal of Organometallic Chemistry, 750, 125-131 (2014)
"Synthesis, analysis of spectroscopic and nonlinear optical properties of the novel compound:(S)-N-benzyl-1-phenyl-5-(thiophen-3-yl)-4-pentyn-2-amine"
Karabacak M, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 97, 556-567 (2012)
"An easy synthetic approach to 1, 2, 3-triazole-fused heterocycles"
Fiandanese V, et al.
Tetrahedron, 66(46), 8846-8853 (2010)
Qing Li et al.
Marine drugs, 16(4) (2018-03-31)
Chitosan is an abundant and renewable polysaccharide, which exhibits attractive bioactivities and natural properties. Improvement such as chemical modification of chitosan is often performed for its potential of providing high bioactivity and good water solubility. A new class of chitosan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service