Skip to Content
Merck
All Photos(1)

Documents

568112

Sigma-Aldrich

Diisopropylzinc solution

1.0 M in toluene

Synonym(s):

Bis(1-methylethyl)zinc, Bis(isopropyl)zinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CH(CH3)2)2
CAS Number:
Molecular Weight:
151.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

1.0 M in toluene

density

0.895 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)[Zn]C(C)C

InChI

1S/2C3H7.Zn/c2*1-3-2;/h2*3H,1-2H3;

InChI key

KDUNMLRPPVCIGP-UHFFFAOYSA-N

Application

Diisopropylzinc is a general reagent used in the organic asymmetric synthesis such as the asymmetric synthesis of [6]?helicene, [5]-helicene and, enantiomerically enriched 5-pyrimidyl alkanol.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Synthesis of Chiral Secondary Alcohols with Isopropyl Substituents by the Catalytic Enantioselective Addition of Diisopropylzinc to Aldehydes Using N, N-Dialkylnorephedrines as Chiral Catalysts.
Soai, Kenso et al.
The Journal of Organic Chemistry, 59(25), 7908-7909 (1994)
Stereoselective preparation and reactions of configurationally defined dialkylzinc compounds.
Boudier, Andreas et al.
Chemistry?A European Journal , 6(15), 2748-2761 (2000)
Highly enantioselective synthesis of organic compound using right-and left-handed helical silica.
Sato, Itaru et al.
Tetrahedron Letters, 44(4), 721-724 (2003)
Asymmetric induction by helical hydrocarbons:[6]-and [5] helicenes.
Sato, Itaru et al.
Angewandte Chemie (International Edition in English), 113(6), 1130-1132 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service