Skip to Content
Merck
All Photos(1)

Key Documents

551899

Sigma-Aldrich

2,3-Diamino-5-bromopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6BrN3
CAS Number:
Molecular Weight:
188.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

155 °C (dec.) (lit.)

functional group

bromo

SMILES string

Nc1cc(Br)cnc1N

InChI

1S/C5H6BrN3/c6-3-1-4(7)5(8)9-2-3/h1-2H,7H2,(H2,8,9)

InChI key

YRGMYJUKFJPNPD-UHFFFAOYSA-N

General description

2,3-Diamino-5-bromopyridine can be prepared from 2-amino-3-nitro-5-bromopyridine via reduction using stannous chloride.

Application

2,3-Diamino-5-bromopyridine may be used in the preparation of the following heterocyclic compounds:
  • 6-bromoimidazo-[b]pyridine
  • 11-bromopyrido[2′,3′:5,6]pyrazino[2,3-f][1,10]phenanthroline
  • 6-bromo-3-(tetrahydro-2H-pyran-2-yl)-3H-imidazo[4,5-b]pyridine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Microwave-Assisted C-2 Direct Alkenylation of Imidazo [4, 5-b] pyridines: Access to Fluorescent Purine Isosteres with Remarkably Large Stokes Shifts.
Baladi T, et al.
European Journal of Organic Chemistry, 2016(14), 2421-2434 (2016)
Metabolite analogs. VIII. Syntheses of some imidazopyridines and pyridotriazoles.
Graboyes H and Day AR.
Journal of the American Chemical Society, 79(24), 6421-6426 (1957)
Re (I) Complexes of Substituted dppz: A Computational and Spectroscopic Study.
van der Salm H, et al.
Inorganic Chemistry, 53(6), 3126-3140 (2014)
Sundaram Ellairaja et al.
Biosensors & bioelectronics, 91, 82-88 (2016-12-20)
Bilirubin, a key biomarker for the jaundice and its clinical diagnosis needs a better analytical tool. A novel and simple fluorescent platform based on (2,2'-((1E,1'E)-((6-bromopyridine-2,3-diyl) bis(azanylylidene)) bis(methanylylidene diphenol) (BAMD) was designed. BAMD showed a remarkable fluorescent intensity with a very
Sheta M Sheta et al.
Dalton transactions (Cambridge, England : 2003), 47(14), 4847-4855 (2018-03-16)
Novel copper metal organic framework nanoparticles Cu-MOF-NPs (C1) were prepared via two simple alternative methods and confirmed by analytical characterization using mass, IR, Raman, XRD spectrum, HR-TEM and TGA-DSC. Mass spectroscopy revealed the molecular ion peak at 647 m/z for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service