Skip to Content
Merck
All Photos(2)

Documents

550256

Sigma-Aldrich

Silver methanesulfonate

Synonym(s):

Silver methylsulfonate, Methanesulfonic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgSO3CH3
CAS Number:
Molecular Weight:
202.97
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: silver
reagent type: catalyst

mp

252-256 °C (lit.)

SMILES string

[Ag+].CS([O-])(=O)=O

InChI

1S/CH4O3S.Ag/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

MLKQJVFHEUORBO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Heterocyclization reactions
  • CO2-mediated rearrangement of propargyl alcohols for the synthesis of a,ß-unsaturated ketones and esters

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsung-Ming Shih et al.
Chemico-biological interactions, 187(1-3), 207-214 (2010-03-13)
This study compared the ability of nine oximes (HI-6, HLö7, MMB-4, TMB-4, carboxime, ICD585, ICD692, ICD3805, and 2-PAM) to reactivate in vivo cholinesterase (ChE) in blood, brain, and peripheral tissues in guinea pigs intoxicated by one of four organophosphorus nerve
Wenyuan Liu et al.
Journal of pharmaceutical and biomedical analysis, 52(4), 603-608 (2010-02-09)
10-O-(N,N-dimethylaminoethyl)-ginkgolide B methanesulfonate (XQ-1H), a novel active derivative of ginkgolide B, is a platelet-activating factor antagonist which is being under clinical trial. Two unknown related impurities were observed in analysis of XQ-1H bulk drug. A scaling up preparative liquid chromatography
Neal W Sach et al.
Organic letters, 14(15), 3886-3889 (2012-07-18)
A general synthesis of aryl ethers from primary and secondary alcohols and aryl mesylates is presented. The reaction proceeds via a sulfonyl-transfer mechanism. In this paper, we compare the sulfonyl transfer reaction to Mitsunobu ether formation. The reaction can be
Benedetto Natalini et al.
Talanta, 85(3), 1392-1397 (2011-08-03)
The successful enantioseparation of five 6-desfluoroquinolones with three polysaccharide-based stationary phases (namely, the cellulose-based Chiralpak IB and the two amylose-based Chiralpak AD-H and Lux Amylose-2) is herein described. The investigated species differ for the nature of substituents and/or the position
Renata Solarska et al.
Nanoscale, 4(5), 1553-1556 (2012-02-01)
Nanostructuring of semiconductor films offers the potential means for producing photoelectrodes with improved minority charge carrier collection. Crucial to the effective operation of the photoelectrode is also the choice of a suitable electrolyte. The behaviour of the nanostructured WO(3) photoanodes

Articles

We are proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service