Skip to Content
Merck
All Photos(2)

Key Documents

544906

Sigma-Aldrich

Zinc oxide

nanopowder, <100 nm particle size

Synonym(s):

Zinc monoxide, Zinc white

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
Molecular Weight:
81.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

~80% Zn basis

form

nanopowder

surface area

10-25 m2/g

particle size

<100 nm

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zinc oxide (ZnO) nanopowder is a metal oxide that is an n-type direct band semiconducting material with a band gap of 3.37 eV. Its properties include high chemical stability, photostability, and high electrochemical coupling coefficient. It is classified as a semiconductor in group II-VI with bond energy of 60 meV, and high thermo-mechanical stability at ambient temperatures.[1][2]

Application

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.
ZnO nanopowder can be used for a variety of applications, such as catalysts, optical materials, UV absorbers, gas sensors and cosmetics.[1][2][3][4]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous doping of aluminum and fluorine on zinc oxide nanopowder using a low-cost soft chemical route
Saravanakumar K, et al.
Materials Letters, 65(14), 2278-2280 (2011)
Javier Orozco-Messana
Nanomaterials (Basel, Switzerland), 10(10) (2020-09-30)
A research study on the application of biomimetic ZnO (from eggshell membranes) as photoanodes in dye-sensitized solar cells (DSSCs) is presented. Biomimetic ZnO powder was produced and characterized. Its surface area, crystallinity, and morphology were analyzed and compared to commercial
Zinc oxide?from synthesis to application: a review
Kolodziejczak-Radzimska A, et al.
Materials, 7(4), 2833-2881 (2014)
"Nanopowder synthesis of zinc oxide via solochemical processing.
Vaezi MR and Sadrnezhaad SK
Materials & Design, 28(2), 515-519 (2007)
Photoluminescence of zinc oxide nanopowder synthesized by a combustion method
Tarwal NL, et al.
Powder Technology, 208(1), 185-188 (2011)

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

See All

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. May I know its band gap value?

    1 answer
    1. The band gap value of this material has not been determined. However, information available in the literature suggests this semiconducting material demonstrates a band gap of 3.37 eV at room temperature. Please refer to the link below to review additional information:
      https://www.sciencedirect.com/science/article/pii/S003810982030661X#:~:text=The%20band%20gap%20of%20bulk,at%20room%20temperature%20%5B6%5D.

      Helpful?

  4. How the ZnO nanoparties product 544906 is synthesised?

    1 answer
    1. The method of synthesis is considered proprietary. The shareable information for this product can be found on the product page, COA, and SDS.

      Helpful?

  5. Please tell me the "Purity" of the ZnO powder, Product number: 544906?

    1 answer
    1. The purity of this product is measured by the Zinc content. The compound is approximately 80% Zn as stated in the 'PROPERTIES' section in 'Assay'.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service