All Photos(2)
About This Item
Linear Formula:
(CH2=CHCH2)2NCH3
CAS Number:
Molecular Weight:
111.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.43 (lit.)
bp
111 °C (lit.)
density
0.789 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(CC=C)CC=C
InChI
1S/C7H13N/c1-4-6-8(3)7-5-2/h4-5H,1-2,6-7H2,3H3
InChI key
WGESLFUSXZBFQF-UHFFFAOYSA-N
General description
Diallylmethylamine is a tertiary amine. It can be prepared from the reaction between aqueous methylamine and allylchloride in the presence of polyglycol-400 (phase transfer catalyst).
Application
Diallylmethylamine may be used in the synthesis of the following:
- diallyldimethylammonium iodide
- diallylethylmethylammonium bromide
- diallylbenzylmethylammonium chloride
- alkyl substituted diallylmethyl quaternary ammonium salt monomers
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
45.0 °F - closed cup
Flash Point(C)
7.2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of Diallylmethylamine by Phase Transfer Catalysis.
Xiao Y, et al.
Chinese Journal of Applied Chemistry / Ying Yong Hua Xue, 21(8), 818-818 (2004)
Base stable pyrrolidinium cations for alkaline anion exchange membrane applications.
Gu F, et al.
Macromolecules, 47(19), 6740-6747 (2014)
Synthesis and Characterization of Diallylmethylalkyl Quaternary Ammonium Salts.
Tao XP and Zhang YJ.
Fine Chemicals / ????, 7, 028-028 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service