Skip to Content
Merck
All Photos(3)

Documents

536180

Sigma-Aldrich

4-Vinylaniline

97%

Synonym(s):

4-Aminostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4NH2
CAS Number:
Molecular Weight:
119.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

refractive index

n20/D 1.626 (lit.)

bp

213-214 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(C=C)cc1

InChI

1S/C8H9N/c1-2-7-3-5-8(9)6-4-7/h2-6H,1,9H2

InChI key

LBSXSAXOLABXMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinylaniline is a vinyl-substituted aromatic amine that is widely used as a crosslinking agent in the production of resins, adhesives, and coatings, enhancing their mechanical properties and chemical resistance. It can be polymerized by methods like radical polymerization, cationic polymerization, or oxidative polymerization to form poly(4-vinylaniline) or P4VA. 4- Vinylaniline can be selectively polymerized or chemically modified to respond to specific analytes or stimuli, allowing the development of conductive polymers for biosensors.

Application

4-Vinylaniline can be used as:

  • A monomer to prepare electrically conductive polyvinylaniline/polyaniline (PVAN/PANI) bilayer for surface modification of bacterial cellulose (BC) nanofibril network. This material can be utilized as an electrochemical biosensor for nerve regenerative medicine.
  • To functionalize carbon nanodots for in vivo fluorometric biosensors, for the detection of mercury and cysteine.
  • As a precursor to prepare conductive and biocompatible hydrogels forwound healing applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinyl-functionalized polyphenolic-carbon dot-based fluorometric turn-off?on biosensor for the dual detection of mercury and cysteine and their in vivo sensing in zebrafish larvae
Buddolla Anantha Lakshmi, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 282, 121685-121685 (2022)
Reactive Coupling of 4-Vinylaniline with Hydrogen-Terminated Si (100) Surfaces for Electroless Metal and ?Synthetic Metal
Xu D, et al.
Langmuir null
Chemical modification of Si (100) surface by consecutive graft polymerization of 4-vinylaniline and aniline
Ji LY, et al.
Reactive functional Polymers, 46(2), 145-156 (2000)
Palladium (II) Schiff base complexes derived from allylamine and vinylaniline
Uh YS, et al.
Bull. Korean Chem. Soc., 25(7), 986-990 (2004)
Synthesis and characterization of styrene grafted carbon nanotube and its polystyrene nanocomposite
Nayak RR, et al.
Eur. Polymer J., 43, 4916-4923 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service