Skip to Content
Merck
All Photos(1)

Documents

521248

Sigma-Aldrich

2-(Methylthio)phenylboronic acid

≥95%

Synonym(s):

2-(Methylthio)benzeneboronic acid, 2-Methylsulfanylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SC6H4B(OH)2
CAS Number:
Molecular Weight:
168.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

77-83 °C (lit.)

SMILES string

CSc1ccccc1B(O)O

InChI

1S/C7H9BO2S/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5,9-10H,1H3

InChI key

QXBWTYBCNFKURT-UHFFFAOYSA-N

Application

Reactant for:
  • Palladium-catalyzed cross-coupling reactions
  • Suzuki-Miyaura coupling reactions

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yae In Cho et al.
Inorganic chemistry, 58(19), 12689-12699 (2019-09-10)
We report syntheses and H2 activation involving model complexes of mono-iron hydrogenase (Hmd) derived from acyl-containing pincer ligand precursors bearing thioether (CNS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service