Skip to Content
Merck
All Photos(6)

Key Documents

520764

Sigma-Aldrich

Palladium(II) acetate

≥99.9% trace metals basis

Synonym(s):

Pd(OAc)2, [Pd(OAc)2]3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
Beilstein:
6086766
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.9% trace metals basis

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: C-H Activation

mp

216.3-223.7 °C (dec.)

SMILES string

CC(O[Pd]OC(C)=O)=O

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Palladium(II) acetate is a homogenous oxidation catalyst. It participates in the activation of alkenic and aromatic compounds towards oxidative inter- and intramolecular nucleophilic reactions. Crystals of palladium(II) acetate have a trimeric structure, having symmetry D3h. Each of the palladium atoms in the crystals are joined to the other two by double acetate bridges. Microencapsulation of palladium(II) acetate in polyurea affords polyurea-encapsulated palladium(II) acetate. It is a versatile heterogeneous catalyst for various phosphine-free cross-coupling reactions. It participates as catalyst in the Heck coupling reaction of pthalides with different alkenes.

Application

Palladium(II) acetate was employed as a catalyst for the following reactions:
  • Formation of allylic acetates. (eq. 1)
  • Hydroselenation of triple bonds. (eq. 2)
  • Heck arylation of alkenes. (eq. 3)
  • Cyclocarbonylation. (eq. 4)
  • Buchwald-Hartwig amination reaction. (eq. 5)
Pd(OAc)<sub>2</sub> catalyst
For small scale and high throughput uses, product is also available as ChemBeads (924377)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile Synthesis of Novel Cyclic Esters of γ-Keto Acid Derivatives by Heck Coupling Reaction.
ShashiKumar ND, et al.
Journal of Heterocyclic Chemistry, 51(S1), E354-E357 (2014)
Wolfe, J. P.; Buchwald, S. L.
The Journal of Organic Chemistry, 62, 1264-1264 (1997)
Palladium (II) Acetate
Grennberg H, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2015)
The crystal structure of Trimeric palladium (II) acetate.
Skapski AC and Smart ML.
Journal of the Chemical Society. Chemical Communications, 11, 658b-6659 (1970)
Palladium(II)-catalyzed C-H bond arylation of electron-deficient arenes at room temperature.
Matthew J Tredwell et al.
Angewandte Chemie (International ed. in English), 50(5), 1076-1079 (2011-01-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service