Skip to Content
Merck
All Photos(1)

Documents

517887

Sigma-Aldrich

trans-4-Phenyl-3-buten-2-one-d10

97 atom % D

Synonym(s):

benzalacetone-d10, benzylideneacetone-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD=CDCOCD3
CAS Number:
Molecular Weight:
156.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

97 atom % D

bp

260-262 °C (lit.)

mp

39-42 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(\[2H])=C(/[2H])C(=O)C([2H])([2H])[2H]

InChI

1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+/i1D3,2D,3D,4D,5D,6D,7D,8D

InChI key

BWHOZHOGCMHOBV-ZLPBFNINSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service