Skip to Content
Merck
All Photos(2)

Documents

499781

Sigma-Aldrich

Benzyl carbazate

97%

Synonym(s):

(Benzyloxycarbonyl)hydrazine, Z-hydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHCO2CH2C6H5
CAS Number:
Molecular Weight:
166.18
Beilstein:
1952982
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

65-68 °C (lit.)

application(s)

peptide synthesis

SMILES string

NNC(=O)OCc1ccccc1

InChI

1S/C8H10N2O2/c9-10-8(11)12-6-7-4-2-1-3-5-7/h1-5H,6,9H2,(H,10,11)

InChI key

RXUBZLMIGSAPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Palanivelu Nithya et al.
Journal of photochemistry and photobiology. B, Biology, 165, 220-231 (2016-11-07)
In the present study, new Schiff base complexes with the composition [M(NCS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service