Skip to Content
Merck
All Photos(1)

Key Documents

498203

Sigma-Aldrich

3,3,3-Trifluoropropionic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2CO2H
CAS Number:
Molecular Weight:
128.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.333 (lit.)

bp

145 °C/746 mmHg (lit.)

mp

9.7 °C (lit.)

density

1.45 g/mL at 25 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)CC(F)(F)F

InChI

1S/C3H3F3O2/c4-3(5,6)1-2(7)8/h1H2,(H,7,8)

InChI key

KSNKQSPJFRQSEI-UHFFFAOYSA-N

Application

3,3,3-Trifluoropropionic acid may be used in the synthesis of:
  • 2,2,2-trifluoroethyl 3,3,3-trifluoroprionate
  • ethyl 3,3,3-trifluoroprionate
  • N-trifluoropropionyl-D-mannosamine
  • 3,3,3-trifluoropropanoyl chloride

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

157 nm Resist Materials: A Progress Report.
Chiba T, et al.
J. Photopolym. Sci. Technol., 13(4), 657-664 (2000)
Bi-phobic cellulose fibers derivatives via surface trifluoropropanoylation.
Cunha AG, et al.
Langmuir, 23(21), 10801-10806 (2007)
Synthesis of ethyl 3,3,3-trifluoropropionate from 2-bromo-3,3,3-trifluoropropene.
Inoue M, et al.
Journal of Fluorine Chemistry, 167, 135-138 (2014)
Yanbin Pan et al.
Carbohydrate research, 339(12), 2091-2100 (2004-07-29)
N-Acetyl-D-neuraminic acid (NeuNAc) aldolase is an important enzyme for the metabolic engineering of cell-surface NeuNAc using chemically modified D-mannosamines. To explore the optimal substrates for this application, eight N-acyl derivatives of D-mannosamine were prepared, and their accessibility to NeuNAc aldolase
Bifan Chen et al.
Analytical chemistry, 91(18), 11661-11669 (2019-08-24)
Antibody-drug conjugates (ADCs) are designed to combine the target specificity of monoclonal antibodies and potent cytotoxin drugs to achieve better therapeutic outcomes. Comprehensive evaluation of the quality attributes of ADCs is critical for drug development but remains challenging due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service