Skip to Content
Merck
All Photos(1)

Documents

49661

Sigma-Aldrich

Methyl 4-pentenoate

≥95.0% (GC)

Synonym(s):

4-Pentenoic acid methyl ester, Allylacetic acid methyl ester, Methyl allylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

refractive index

n20/D 1.415
n20/D 1.415

SMILES string

COC(=O)CCC=C

InChI

1S/C6H10O2/c1-3-4-5-6(7)8-2/h3H,1,4-5H2,2H3

InChI key

SHCSFZHSNSGTOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 4-pentenoate is an unsaturated methyl ester that is prepared by the esterification of 1-pentenoic acid with methanol. It undergoes amidation with formamide via acetone-initiated photochemical reaction to form a 1:1 adduct. The metathesis of methyl 4-pentenoate with different Mo(VI)alkylidene complexes have been reported. It acts as a chain transfer agent during the polymerization of exo,exo-5,6-bis(methoxymethyl)-7-oxabicyclo[2.2.1]hept-2-ene using RuII(H20)6(tos)2 (tos =p-toluenesulfonate) as a catalyst.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Light-Induced Amidation of Terminal Olefins1.
Elad D and Rokach J.
The Journal of Organic Chemistry, 29(7), 1855-1859 (1964)
Coupling of terminal olefins by molybdenum (VI) imido alkylidene complexes.
Fox HH, et al.
Organometallics, 13(2), 635-639 (1994)
Chain transfer during the aqueous ring-opening metathesis polymerization of 7-oxanorbornene derivatives.
France MB, et al.
Macromolecules, 26(18), 4742-4747 (1993)
Majd Al-Naji et al.
ChemSusChem, 12(12), 2628-2636 (2019-04-18)
The need for more sustainable products and processes has led to the use of new methodologies with low carbon footprints. In this work, an efficient tandem process is demonstrated for the liquid-phase catalytic upgrading of lignocellulosic biomass-derived γ-valerolactone (GVL) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service