Unfortunately, the molecular weight and other physical properties of this material have not been determined. This product is a white powder. Features such as boiling point would not be applicable. The monomeric Ethylene starting material has a molecular weight of 30.04 g/mol.
Recommended Products
isotopic purity
99 atom % 13C
form
solid
mp
130-145 °C (lit.)
mass shift
M+2 (per repeating unit)
storage temp.
room temp
SMILES string
[X][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][X]
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Hello, What is the average molecular weight of this polyethylene? Can I get more information about this polyethylene such as boiling temperature, molecular weight..?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service