491268
Isoamyl nitrite-15N
98 atom % 15N, 97% (CP)
Synonym(s):
15N Labeled isoamyl nitrite, Isopentyl nitrite-15N
About This Item
Recommended Products
isotopic purity
98 atom % 15N
Assay
97% (CP)
refractive index
n20/D 1.386 (lit.)
bp
99 °C (lit.)
density
0.879 g/mL at 25 °C (lit.)
mass shift
M+1
SMILES string
CC(C)CCO[15N]=O
InChI
1S/C5H11NO2/c1-5(2)3-4-8-6-7/h5H,3-4H2,1-2H3/i6+1
InChI key
OWFXIOWLTKNBAP-PTQBSOBMSA-N
Related Categories
Application
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1
WGK
WGK 3
Flash Point(F)
closed cup
Flash Point(C)
closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service