Skip to Content
Merck
All Photos(1)

Key Documents

491187

Sigma-Aldrich

17β-Estradiol-16,16,17-d3

98 atom % D, 99% (CP)

Synonym(s):

β-Estradiol-16,16,17-d3, 1,3,5-Estratriene-3,17β-diol-16,16,17-d3, 3,17β-Dihydroxy-1,3,5(10)-estratriene-16,16,17-d3, Deuterated 17β-estradiol, Dihydrofolliculin-16,16,17-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D3H21O2
CAS Number:
Molecular Weight:
275.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

178-179 °C (lit.)

mass shift

M+3

SMILES string

[2H]C1([2H])C[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]2(C)[C@@]1([2H])O

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i7D2,17D

InChI key

VOXZDWNPVJITMN-SPGJGCHISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Ying Ru et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 250, 110492-110492 (2020-09-06)
Growth hormone (GH) is the most important endocrine factor to regulate somatic growth. Spotted scat (Scatophagus argus) is a famous marine aquaculture species in China with a typical sexual growth dimorphism in which females grow faster and larger than males.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service