Skip to Content
Merck
All Photos(1)

Documents

488895

Sigma-Aldrich

Sodium DL-3-hydroxybutyrate-1,3-13C2

99 atom % 13C

Synonym(s):

DL-3-Hydroxybutyric acid-1,3-13C2 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CH(OH)CH213CO2Na
CAS Number:
Molecular Weight:
128.07
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

161-167 °C (lit.)

mass shift

M+2

storage temp.

2-8°C

SMILES string

[Na+].C[13CH](O)C[13C]([O-])=O

InChI

1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/i3+1,4+1;

InChI key

NBPUSGBJDWCHKC-ZPGUIXIESA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jordan M Wilkins et al.
Aging, 12(14), 15134-15156 (2020-07-09)
Multiple sclerosis (MS) is a central nervous system inflammatory demyelinating disease and the most common cause of non-traumatic disability in young adults. Despite progress in the treatment of the active relapsing disease, therapeutic options targeting irreversible progressive decline remain limited.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service