Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
Cl2C6D3OH
CAS Number:
Molecular Weight:
166.02
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Isotopic purity:
98 atom % D
Assay:
98% (CP)
Mass shift:
M+3
Form:
solid
Pricing and availability is not currently available.
isotopic purity
98 atom % D
Quality Segment
assay
98% (CP)
form
solid
bp
209-210 °C (lit.)
mp
42-43 °C (lit.)
mass shift
M+3
SMILES string
[2H]c1c([2H])c(Cl)c([2H])c(Cl)c1O
InChI
1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H/i1D,2D,3D
InChI key
HFZWRUODUSTPEG-CBYSEHNBSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
1 of 1
This Item | |||
|---|---|---|---|
| assay 98% (CP) | assay 99% (CP) | assay - | assay ≥99% (CP) |
| isotopic purity 98 atom % D | isotopic purity 98 atom % D | isotopic purity 98 atom % D | isotopic purity ≥98 atom % D |
| mass shift M+3 | mass shift M+3 | mass shift M+4 | mass shift M+3 |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form solid | form solid | form solid | form solid |
| mp 42-43 °C (lit.) | mp 22-23 °C (lit.) | mp 43-45 °C (lit.) | mp 40-42 °C (lit.) |
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
237.2 °F - closed cup
flash_point_c
114 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






