Skip to Content
Merck
All Photos(1)

Key Documents

485306

Sigma-Aldrich

Ammonium-14N2 sulfate solution

40 wt. % in H2O, 99.99 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(14NH4)2SO4/H2O soln.
CAS Number:
Molecular Weight:
132.13
EC Number:
MDL number:
UNSPSC Code:
12352314
PubChem Substance ID:

description

15N-depleted

Quality Level

isotopic purity

99.99 atom % 14N

form

solid

concentration

40 wt. % in H2O

refractive index

n20/D 1.396

density

1.23 g/mL at 25 °C

mass shift

depleted

SMILES string

[14NH3].[14NH3].OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+0;

InChI key

BFNBIHQBYMNNAN-ONPSQTMSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)
Kazunori Sugiura et al.
Biochemical and biophysical research communications, 457(3), 242-248 (2015-01-17)
Intracellular redox state is a critical factor for fundamental cellular functions, including regulation of the activities of various metabolic enzymes as well as ROS production and elimination. Genetically-encoded fluorescent redox sensors, such as roGFP (Hanson, G. T., et al. (2004))
Adam Kawałek et al.
Aging, 5(1), 67-83 (2013-02-22)
We studied the role of peroxisomal catalase in chronological aging of the yeastHansenula polymorpha in relation to various growth substrates. Catalase-deficient (cat) cells showed a similar chronological life span (CLS) relative to the wild-type control upon growth on carbon and
Xiangyao Liu et al.
Carbohydrate polymers, 92(1), 659-661 (2012-12-12)
Cellulose fibers were engineered by in situ oxidative polymerization of aniline using ammonium persulfate as oxidant/initiator. The polyaniline/cellulose fiber composite was used for the treatment of Cr(VI)-contaminated water, and its effect was evaluated. Under the conditions studied, the composite exhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service