Skip to Content
Merck
All Photos(2)

Key Documents

481181

Sigma-Aldrich

Silver(I) sulfadiazine

98%

Synonym(s):

Silver sulfadiazine, Sulfadiazine silver

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9AgN4O2S
CAS Number:
Molecular Weight:
357.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder

Quality Level

Assay

98%

form

powder

mp

285 °C (dec.) (lit.)

SMILES string

Nc1ccc(cc1)S(=O)(=O)N([Ag])c2ncccn2

InChI

1S/C10H9N4O2S.Ag/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10;/h1-7H,11H2;/q-1;+1

InChI key

UEJSSZHHYBHCEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silver(I) sulfadiazine (AgSD) is popularly used for treating second-degree burn wounds as it acts as an antimicrobial agent and prevents infections. Silver sulfadiazine-impregnated polyurethane (PU) foam, chitosan/chondroitin sulfate films may be used for wound dressing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver sulfadiazine loaded chitosan/chondroitin sulfate films for a potential wound dressing application
Fajardo AR, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 33(2), 588-595 (2013)
Vincenzina Monzillo et al.
The new microbiologica, 35(2), 175-182 (2012-06-19)
The aim of this study was to compare the in vitro activity and the impact on bacterial adhesion of two different catheters, one impregnated with chlorhexidine-silver sulfadiazine (C-SS) and the other not impregnated with antibacterial agents. The antimicrobial coating prevented
[Modern possibilities of the local treatment of children with burn wounds].
L I Budkevich et al.
Khirurgiia, (10)(10), 48-51 (2012-12-14)
Steven L Percival et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 20(5), 647-657 (2012-09-19)
Both chronic and acute dermal wounds are susceptible to infection due to sterile loss of the innate barrier function of the skin and dermal appendages, facilitating the development of microbial communities, referred to as biofilms, within the wound environment. Microbial
Chia-Yu Chu et al.
PloS one, 7(6), e38360-e38360 (2012-06-14)
Silver nanoparticles supported on nanoscale silicate platelets (AgNP/NSP) possess interesting properties, including a large surface area and high biocide effectiveness. The nanohybrid of AgNP/NSP at a weight ratio 7/93 contains 5-nm Ag particles supported on the surface of platelets with

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service