Skip to Content
Merck
All Photos(1)

Documents

480754

Sigma-Aldrich

tert-Butyl diazoacetate

Synonym(s):

(tert-Butoxycarbonyl)diazomethane, 1,1-Dimethylethyl diazoacetate, 2-Diazoacetic acid tert-butyl ester, 2-Diazonio-1-[(2-methylpropan-2-yl)oxy]ethenolate, Diazoacetic acid 1,1-dimethylethyl ester, tert-Butyl α-diazoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N2CHCO2C(CH3)3
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

impurities

≤15% dichloromethane

Quality Level

refractive index

n20/D 1.453 (lit.)

bp

51-53 °C/12 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)C=[N+]=[N-]

InChI

1S/C6H10N2O2/c1-6(2,3)10-5(9)4-8-7/h4H,1-3H3

InChI key

JBVSBLLOZVDAAZ-UHFFFAOYSA-N

General description

tert-Butyl diazoacetate can be used for numerous organic transformations, including cyclopropanation, insertion, and aziridine-forming reactions. 

Application

tert-Butyl diazoacetate may be used for the preparation of tert-butyl 2-(4-chlorophenyl)cyclopropane-1-carboxylate with high enantioselectivity and trans-cis selectivity via cyclopropanation with 4-chlorostyrene in the presence of a chiral (salen)cobalt(III) bromide complex as a catalyst.
It is a key starting material in the total synthesis of manzacidin A. It can also be used in the synthesis of diastero- and enantioselective synthesis of 2-vinylcyclopropa[b]indolines from 2-vinylindoles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective 1, 3-dipolar cycloaddition reaction between diazoacetates and α-substituted acroleins: total synthesis of manzacidin A.
Kano T, et al.
Journal of the American Chemical Society, 128(7), 2174-2175 (2006)
[Copper (I)(Pyridine-Containing Ligand)] Catalyzed Regio-and Steroselective Synthesis of 2-Vinylcyclopropa [b] indolines from 2-Vinylindoles.
Pirovano V, et al.
Organic Letters, 20(2), 405-408 (2018)
Highly enantioselective cyclopropanation of styrene derivatives using Co (III)-salen complex as a catalyst.
Fukuda T & Katsuki T
Tetrahedron, 53(21), 7201-7208 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service